Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATAGGTCTAACTTGGGTGTTTC[A/C]AAAAATGTCAAGTAACGAACACACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf159 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C14orf159 - chromosome 14 open reading frame 159 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102366.2 | Intron | NP_001095836.1 | ||||
NM_001102367.2 | Intron | NP_001095837.1 | ||||
NM_001102368.2 | Intron | NP_001095838.1 | ||||
NM_001102369.2 | Intron | NP_001095839.1 | ||||
NM_001286470.1 | Intron | NP_001273399.1 | ||||
NM_001286471.1 | Intron | NP_001273400.1 | ||||
NM_001286472.1 | Intron | NP_001273401.1 | ||||
NM_001286473.1 | Intron | NP_001273402.1 | ||||
NM_024952.7 | Intron | NP_079228.4 |
SNORA11B - small nucleolar RNA, H/ACA box 11B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |