Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605004 MIM: 603065 | |||||||||||||||||||||||
Literature Links: |
GSK3B PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GSK3B - glycogen synthase kinase 3 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146156.1 | Intron | NP_001139628.1 | ||||
NM_002093.3 | Intron | NP_002084.2 | ||||
XM_006713610.2 | Intron | XP_006713673.1 | ||||
XM_006713611.2 | Intron | XP_006713674.1 |
NR1I2 - nuclear receptor subfamily 1 group I member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |