Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610487 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KHDRBS2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KHDRBS2 - KH RNA binding domain containing, signal transduction associated 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152688.3 | Intron | NP_689901.2 | ||||
XM_011535515.2 | Intron | XP_011533817.1 | ||||
XM_011535516.1 | Intron | XP_011533818.1 | ||||
XM_011535517.1 | Intron | XP_011533819.1 | ||||
XM_011535519.1 | Intron | XP_011533821.1 | ||||
XM_011535520.1 | Intron | XP_011533822.1 | ||||
XM_017010334.1 | Intron | XP_016865823.1 | ||||
XM_017010335.1 | Intron | XP_016865824.1 | ||||
XM_017010336.1 | Intron | XP_016865825.1 | ||||
XM_017010337.1 | Intron | XP_016865826.1 | ||||
XM_017010338.1 | Intron | XP_016865827.1 | ||||
XM_017010339.1 | Intron | XP_016865828.1 | ||||
XM_017010340.1 | Intron | XP_016865829.1 | ||||
XM_017010341.1 | Intron | XP_016865830.1 | ||||
XM_017010342.1 | Intron | XP_016865831.1 | ||||
XM_017010343.1 | Intron | XP_016865832.1 | ||||
XM_017010344.1 | Intron | XP_016865833.1 | ||||
XM_017010345.1 | Intron | XP_016865834.1 | ||||
XM_017010346.1 | Intron | XP_016865835.1 | ||||
XM_017010347.1 | Intron | XP_016865836.1 | ||||
XM_017010348.1 | Intron | XP_016865837.1 |
Set Membership: |
HapMap |