Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGTTAACTTTTCAACACAGTGAC[A/C]GAATGAAAAGAGCAGGTTGTATTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610191 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AQP4-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AQP4-AS1 - AQP4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CHST9 - carbohydrate sulfotransferase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256316.1 | 8755 | Intron | NP_001243245.1 | |||
NM_031422.5 | 8755 | Intron | NP_113610.2 | |||
XM_005258362.3 | 8755 | UTR 3 | XP_005258419.1 | |||
XM_006722555.3 | 8755 | Intron | XP_006722618.1 | |||
XM_017026033.1 | 8755 | UTR 3 | XP_016881522.1 | |||
XM_017026034.1 | 8755 | UTR 3 | XP_016881523.1 |
Set Membership: |
HapMap |