Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
53 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614214 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
AFR
|
Chinese - Not Available | JPT (Japanese)
|
||||||
SAS - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
KLHL6 - kelch like family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130446.2 | 2557 | UTR 3 | NP_569713.2 | |||
XM_011513273.2 | 2557 | UTR 3 | XP_011511575.1 | |||
XM_011513274.2 | 2557 | Intron | XP_011511576.1 |
Set Membership: |
HapMap |