Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCTCTGTGCCCATGTATGCTCA[C/T]TCCTTGCATTTGAAAGTAAACTATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612787 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PUS10 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
PUS10 - pseudouridylate synthase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322123.1 | Intron | NP_001309052.1 | ||||
NM_001322124.1 | Intron | NP_001309053.1 | ||||
NM_001322127.1 | Intron | NP_001309056.1 | ||||
NM_144709.3 | Intron | NP_653310.2 | ||||
XM_011532568.2 | Intron | XP_011530870.1 | ||||
XM_011532570.1 | Intron | XP_011530872.1 | ||||
XM_011532571.1 | Intron | XP_011530873.1 | ||||
XM_011532572.1 | Intron | XP_011530874.1 | ||||
XM_011532573.2 | Intron | XP_011530875.1 | ||||
XM_011532574.2 | Intron | XP_011530876.1 | ||||
XM_011532576.2 | Intron | XP_011530878.1 | ||||
XM_011532578.1 | Intron | XP_011530880.1 | ||||
XM_017003428.1 | Intron | XP_016858917.1 | ||||
XM_017003429.1 | Intron | XP_016858918.1 |