Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300451 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EDA PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EDA - ectodysplasin A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005609.1 | 4898 | Intron | NP_001005609.1 | |||
NM_001005610.3 | 4898 | Intron | NP_001005610.2 | |||
NM_001005612.2 | 4898 | Intron | NP_001005612.2 | |||
NM_001005613.3 | 4898 | Intron | NP_001005613.1 | |||
NM_001399.4 | 4898 | Intron | NP_001390.1 | |||
XM_006724630.2 | 4898 | Intron | XP_006724693.1 | |||
XM_011530885.2 | 4898 | Intron | XP_011529187.1 | |||
XM_017029336.1 | 4898 | Intron | XP_016884825.1 | |||
XM_017029337.1 | 4898 | UTR 3 | XP_016884826.1 |