Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142370 | ||||||||||||||||||||
Literature Links: |
HCK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HCK - HCK proto-oncogene, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TM9SF4 - transmembrane 9 superfamily member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014742.3 | 163 | UTR 5 | NP_055557.2 | |||
XM_005260622.4 | 163 | Intron | XP_005260679.1 | |||
XM_017028154.1 | 163 | Intron | XP_016883643.1 | |||
XM_017028155.1 | 163 | Intron | XP_016883644.1 | |||
XM_017028156.1 | 163 | Intron | XP_016883645.1 | |||
XM_017028157.1 | 163 | Intron | XP_016883646.1 | |||
XM_017028158.1 | 163 | Intron | XP_016883647.1 | |||
XM_017028159.1 | 163 | Intron | XP_016883648.1 | |||
XM_017028160.1 | 163 | Intron | XP_016883649.1 | |||
XM_017028161.1 | 163 | Intron | XP_016883650.1 |