Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACATGAAGACCAAAAGATGAGGAC[C/T]TCTTGGTGTCATAAGTGTAGGAAGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605437 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRKCH PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
PRKCH - protein kinase C eta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006255.4 | Intron | NP_006246.2 | ||||
XM_011536954.2 | Intron | XP_011535256.1 | ||||
XM_011536955.1 | Intron | XP_011535257.1 | ||||
XM_011536956.1 | Intron | XP_011535258.1 | ||||
XM_011536957.1 | Intron | XP_011535259.1 | ||||
XM_017021458.1 | Intron | XP_016876947.1 | ||||
XM_017021459.1 | Intron | XP_016876948.1 |
Set Membership: |
HapMap |