Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTTTGTGTCCCTCAACCAGGAAT[C/T]CAAAAAGAGCCACATAAATGCAAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 137140 | ||||||||||||||||||||
Literature Links: |
GABRA2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GABRA2 - gamma-aminobutyric acid type A receptor alpha2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005248080.3 | 4847 | UTR 3 | XP_005248137.1 | |||
XM_011513675.1 | 4847 | UTR 3 | XP_011511977.1 | |||
XM_011513676.1 | 4847 | UTR 3 | XP_011511978.1 | |||
XM_017007982.1 | 4847 | UTR 3 | XP_016863471.1 | |||
XM_017007983.1 | 4847 | UTR 3 | XP_016863472.1 |