Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATACACTAACATTTATTTTTCCA[A/G]TCAATTCATTGCAAATCAGTTTACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605240 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCL28 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CCL28 - C-C motif chemokine ligand 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301873.1 | Intron | NP_001288802.1 | ||||
NM_001301874.1 | Intron | NP_001288803.1 | ||||
NM_001301875.1 | Intron | NP_001288804.1 | ||||
NM_148672.3 | Intron | NP_683513.1 |
Set Membership: |
HapMap |