Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
54 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611445 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf105 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C1orf105 - chromosome 1 open reading frame 105 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300760.1 | 1499 | Intron | NP_001287689.1 | |||
NM_139240.3 | 1499 | Intron | NP_640333.3 | |||
XM_005245608.2 | 1499 | Intron | XP_005245665.1 | |||
XM_005245610.3 | 1499 | Intron | XP_005245667.1 | |||
XM_006711630.2 | 1499 | Intron | XP_006711693.1 | |||
XM_011510152.2 | 1499 | UTR 5 | XP_011508454.1 | |||
XM_011510153.2 | 1499 | UTR 5 | XP_011508455.1 | |||
XM_011510154.2 | 1499 | UTR 5 | XP_011508456.1 | |||
XM_011510155.2 | 1499 | UTR 5 | XP_011508457.1 | |||
XM_011510156.2 | 1499 | UTR 5 | XP_011508458.1 | |||
XM_017002808.1 | 1499 | Intron | XP_016858297.1 |
DNM3 - dynamin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |