Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
39 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602336 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ROR1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ROR1 - receptor tyrosine kinase like orphan receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083592.1 | Intron | NP_001077061.1 | ||||
NM_005012.3 | Intron | NP_005003.2 | ||||
XM_011541526.1 | Intron | XP_011539828.1 | ||||
XM_017001376.1 | Intron | XP_016856865.1 | ||||
XM_017001377.1 | Intron | XP_016856866.1 |
Set Membership: |
HapMap |