Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611776 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL32 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KLHL32 - kelch like family member 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286250.1 | Intron | NP_001273179.1 | ||||
NM_001286251.1 | Intron | NP_001273180.1 | ||||
NM_001286252.1 | Intron | NP_001273181.1 | ||||
NM_001286254.2 | Intron | NP_001273183.1 | ||||
NM_001323252.1 | Intron | NP_001310181.1 | ||||
NM_001323253.1 | Intron | NP_001310182.1 | ||||
NM_001323254.1 | Intron | NP_001310183.1 | ||||
NM_001323255.1 | Intron | NP_001310184.1 | ||||
NM_001323256.1 | Intron | NP_001310185.1 | ||||
NM_001323257.1 | Intron | NP_001310186.1 | ||||
NM_001323258.1 | Intron | NP_001310187.1 | ||||
NM_001323259.1 | Intron | NP_001310188.1 | ||||
NM_001323260.1 | Intron | NP_001310189.1 | ||||
NM_001323261.1 | Intron | NP_001310190.1 | ||||
NM_001323262.1 | Intron | NP_001310191.1 | ||||
NM_001323263.1 | Intron | NP_001310192.1 | ||||
NM_001323264.1 | Intron | NP_001310193.1 | ||||
NM_052904.3 | Intron | NP_443136.2 | ||||
XM_005266813.4 | Intron | XP_005266870.1 | ||||
XM_017010228.1 | Intron | XP_016865717.1 | ||||
XM_017010229.1 | Intron | XP_016865718.1 |
NDUFAF4 - NADH:ubiquinone oxidoreductase complex assembly factor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |