Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 142370 | |||||||||||||||||||||||
Literature Links: |
HCK PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
HCK - HCK proto-oncogene, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172129.1 | Intron | NP_001165600.1 | ||||
NM_001172130.1 | Intron | NP_001165601.1 | ||||
NM_001172131.1 | Intron | NP_001165602.1 | ||||
NM_001172132.1 | Intron | NP_001165603.1 | ||||
NM_001172133.1 | Intron | NP_001165604.1 | ||||
NM_002110.3 | Intron | NP_002101.2 |