Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603148 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATF3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATF3 - activating transcription factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030287.3 | Intron | NP_001025458.1 | ||||
NM_001040619.2 | Intron | NP_001035709.1 | ||||
NM_001206484.2 | Intron | NP_001193413.2 | ||||
NM_001206486.2 | Intron | NP_001193415.1 | ||||
NM_001206488.2 | Intron | NP_001193417.2 | ||||
NM_001674.3 | Intron | NP_001665.1 | ||||
XM_005273146.1 | Intron | XP_005273203.1 | ||||
XM_011509579.1 | Intron | XP_011507881.1 |
Set Membership: |
HapMap |