Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAACAAAATAGGTATAGATGGTGA[A/G]TAAAGGACAAAAATAACTAGCATTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF347 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZNF347 - zinc finger protein 347 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172674.1 | 8000 | Intron | NP_001166145.1 | |||
NM_001172675.1 | 8000 | Intron | NP_001166146.1 | |||
NM_032584.2 | 8000 | Intron | NP_115973.2 | |||
XM_005259335.4 | 8000 | UTR 3 | XP_005259392.1 | |||
XM_005259336.4 | 8000 | UTR 3 | XP_005259393.1 | |||
XM_017027384.1 | 8000 | UTR 3 | XP_016882873.1 | |||
XM_017027385.1 | 8000 | UTR 3 | XP_016882874.1 | |||
XM_017027386.1 | 8000 | UTR 3 | XP_016882875.1 |
ZNF415 - zinc finger protein 415 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |