Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAGGAAGGTTGAGAGAGAGGTGC[G/T]GGGGTCTGCGTCTATCTCTGTCGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603615 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAD54L PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RAD54L - RAD54-like (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142548.1 | 112 | Intron | NP_001136020.1 | |||
NM_003579.3 | 112 | UTR 5 | NP_003570.2 | |||
XM_006710975.3 | 112 | Intron | XP_006711038.1 | |||
XM_011542299.2 | 112 | Intron | XP_011540601.1 | |||
XM_011542300.2 | 112 | Intron | XP_011540602.1 |