Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGGCCGGCGGGGGGCAGTAGGT[G/T]TCCAGTTCTGGCTGGGTGGTGTCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH3TC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SH3TC1 - SH3 domain and tetratricopeptide repeats 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318480.1 | Intron | NP_001305409.1 | ||||
NM_018986.4 | Intron | NP_061859.3 | ||||
XM_006713889.2 | Intron | XP_006713952.1 | ||||
XM_006713891.3 | Intron | XP_006713954.1 | ||||
XM_011513488.1 | Intron | XP_011511790.1 | ||||
XM_011513489.2 | Intron | XP_011511791.1 | ||||
XM_017008300.1 | Intron | XP_016863789.1 | ||||
XM_017008301.1 | Intron | XP_016863790.1 | ||||
XM_017008302.1 | Intron | XP_016863791.1 |