Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USPL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
USPL1 - ubiquitin specific peptidase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321532.1 | Intron | NP_001308461.1 | ||||
NM_001321533.1 | Intron | NP_001308462.1 | ||||
NM_001321534.1 | Intron | NP_001308463.1 | ||||
NM_005800.4 | Intron | NP_005791.3 | ||||
XM_005266213.3 | Intron | XP_005266270.1 | ||||
XM_006719751.3 | Intron | XP_006719814.1 | ||||
XM_017020314.1 | Intron | XP_016875803.1 | ||||
XM_017020315.1 | Intron | XP_016875804.1 | ||||
XM_017020316.1 | Intron | XP_016875805.1 |
Set Membership: |
HapMap |