Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGAATATGTGTATGTGATTTAC[C/G]TATGACTATGCCCTAAAGAAAAGCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608140 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUP35 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NUP35 - nucleoporin 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287584.1 | 197 | Intron | NP_001274513.1 | |||
NM_001287585.1 | 197 | Intron | NP_001274514.1 | |||
NM_138285.4 | 197 | Intron | NP_612142.2 | |||
XM_006712254.2 | 197 | Intron | XP_006712317.1 | |||
XM_011510576.2 | 197 | Intron | XP_011508878.1 | |||
XM_011510577.2 | 197 | Intron | XP_011508879.1 | |||
XM_017003308.1 | 197 | UTR 5 | XP_016858797.1 |