Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616049 | ||||||||||||||||||||
Literature Links: |
PABPC1L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PABPC1L - poly(A) binding protein cytoplasmic 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001124756.2 | 898 | Intron | NP_001118228.1 |
TOMM34 - translocase of outer mitochondrial membrane 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006809.4 | 898 | Silent Mutation | AAA,AAG | K,K 275 | NP_006800.2 | |
XM_005260254.2 | 898 | Silent Mutation | AAA,AAG | K,K 173 | XP_005260311.1 | |
XM_011528501.1 | 898 | Silent Mutation | AAA,AAG | K,K 249 | XP_011526803.1 | |
XM_017027600.1 | 898 | UTR 3 | XP_016883089.1 |