Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGTCATAGGGATTAACAGCTTGG[C/T]CAGGGCTAAGCACAAATCCAGGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610055 | ||||||||||||||||||||
Literature Links: |
C19orf57 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C19orf57 - chromosome 19 open reading frame 57 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024323.3 | Intron | NP_077299.3 | ||||
XM_006722885.3 | Intron | XP_006722948.2 | ||||
XM_011528286.2 | Intron | XP_011526588.1 | ||||
XM_011528287.2 | Intron | XP_011526589.1 | ||||
XM_017027300.1 | Intron | XP_016882789.1 | ||||
XM_017027301.1 | Intron | XP_016882790.1 |
CC2D1A - coiled-coil and C2 domain containing 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017721.4 | Intron | NP_060191.3 | ||||
XM_005259972.2 | Intron | XP_005260029.1 | ||||
XM_005259973.2 | Intron | XP_005260030.1 | ||||
XM_005259974.2 | Intron | XP_005260031.1 | ||||
XM_005259975.2 | Intron | XP_005260032.1 |