Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTACCTAGAAAAATATACACATA[C/T]GGTAAAGATTTGCTGGCCTGCTGGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611414 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf44 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C19orf44 - chromosome 19 open reading frame 44 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288834.1 | Intron | NP_001275763.1 | ||||
NM_032207.3 | Intron | NP_115583.1 | ||||
XM_006722920.3 | Intron | XP_006722983.1 | ||||
XM_011528354.2 | Intron | XP_011526656.1 | ||||
XM_017027350.1 | Intron | XP_016882839.1 |
CALR3 - calreticulin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |