Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAATGCCTATTTCATTCTTGTTA[A/G]TTTCCATGCTCCCATGCTCATGGTA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 603968 MIM: 607845 | |||||||||||||||||||||||
Literature Links: |
POLH PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
POLH - DNA polymerase eta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291969.1 | Intron | NP_001278898.1 | ||||
NM_001291970.1 | Intron | NP_001278899.1 | ||||
NM_006502.2 | Intron | NP_006493.1 | ||||
XM_005249186.3 | Intron | XP_005249243.1 | ||||
XM_011514698.2 | Intron | XP_011513000.1 |
XPO5 - exportin 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |