Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614795 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGPAT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
AGPAT4 - 1-acylglycerol-3-phosphate O-acyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020133.2 | 7692 | UTR 3 | NP_064518.1 | |||
XM_005267053.3 | 7692 | UTR 3 | XP_005267110.1 | |||
XM_006715514.3 | 7692 | UTR 3 | XP_006715577.1 | |||
XM_017011059.1 | 7692 | UTR 3 | XP_016866548.1 |
Set Membership: |
HapMap |