Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGATGAGGTAGGGTGTAGAGTGT[A/G]TTAGTTATCTATTACTGCATGATAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613529 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP152 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CEP152 - centrosomal protein 152 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001194998.1 | Intron | NP_001181927.1 | ||||
NM_014985.3 | Intron | NP_055800.2 | ||||
XM_006720437.3 | Intron | XP_006720500.1 | ||||
XM_011521373.2 | Intron | XP_011519675.1 | ||||
XM_011521374.2 | Intron | XP_011519676.1 | ||||
XM_011521375.2 | Intron | XP_011519677.1 | ||||
XM_011521378.2 | Intron | XP_011519680.1 | ||||
XM_011521379.2 | Intron | XP_011519681.1 | ||||
XM_011521381.2 | Intron | XP_011519683.1 | ||||
XM_017022014.1 | Intron | XP_016877503.1 | ||||
XM_017022015.1 | Intron | XP_016877504.1 | ||||
XM_017022016.1 | Intron | XP_016877505.1 |
Set Membership: |
HapMap |