Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608725 | ||||||||||||||||||||
Literature Links: |
PHACTR3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PHACTR3 - phosphatase and actin regulator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199505.1 | Intron | NP_001186434.1 | ||||
NM_001199506.1 | Intron | NP_001186435.1 | ||||
NM_001281507.1 | Intron | NP_001268436.1 | ||||
NM_080672.4 | Intron | NP_542403.1 | ||||
NM_183244.1 | Intron | NP_899067.1 | ||||
NM_183246.1 | Intron | NP_899069.1 |