Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCAGAGCTTTCCTAGCTGGGCA[A/G]GATTCCTGCATCTGCCCTCTGTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM78B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM78B - family with sequence similarity 78 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017961.4 | 4125 | Intron | NP_001017961.1 | |||
NM_001320302.1 | 4125 | UTR 3 | NP_001307231.1 | |||
XM_017000417.1 | 4125 | Intron | XP_016855906.1 | |||
XM_017000418.1 | 4125 | Intron | XP_016855907.1 |
LOC105371586 - uncharacterized LOC105371586 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |