Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACCCTCTTATTCTTCTGCTTTGT[A/G]ATCCTTCTCAGGCCCTTCCTGGGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615130 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT11 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GALNT11 - polypeptide N-acetylgalactosaminyltransferase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304514.1 | Intron | NP_001291443.1 | ||||
NM_022087.3 | Intron | NP_071370.2 | ||||
XM_006716082.2 | Intron | XP_006716145.1 | ||||
XM_006716083.2 | Intron | XP_006716146.1 | ||||
XM_006716084.2 | Intron | XP_006716147.1 | ||||
XM_017012499.1 | Intron | XP_016867988.1 | ||||
XM_017012500.1 | Intron | XP_016867989.1 |
Set Membership: |
HapMap |