Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
51 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602274 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GALNT2 - polypeptide N-acetylgalactosaminyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291866.1 | Intron | NP_001278795.1 | ||||
NM_004481.4 | Intron | NP_004472.1 | ||||
XM_017000963.1 | Intron | XP_016856452.1 | ||||
XM_017000964.1 | Intron | XP_016856453.1 | ||||
XM_017000965.1 | Intron | XP_016856454.1 | ||||
XM_017000966.1 | Intron | XP_016856455.1 |
Set Membership: |
HapMap |