Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTACAAGGAAACTCAACTGGCC[A/G]AAATGAATAGTGGGGGAATGTGAGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IRGQ PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IRGQ - immunity related GTPase Q | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007561.2 | 4376 | UTR 3 | NP_001007562.1 | |||
XM_005258515.3 | 4376 | UTR 3 | XP_005258572.1 |
PINLYP - phospholipase A2 inhibitor and LY6/PLAUR domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF576 - zinc finger protein 576 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |