Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610152 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPM PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CENPM - centromere protein M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002876.2 | Intron | NP_001002876.1 | ||||
NM_001110215.2 | Intron | NP_001103685.1 | ||||
NM_001304370.1 | Intron | NP_001291299.1 | ||||
NM_001304371.1 | Intron | NP_001291300.1 | ||||
NM_001304372.1 | Intron | NP_001291301.1 | ||||
NM_001304373.1 | Intron | NP_001291302.1 | ||||
NM_024053.4 | Intron | NP_076958.1 | ||||
XM_011530368.2 | Intron | XP_011528670.1 |
LINC00634 - long intergenic non-protein coding RNA 634 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |