Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGGTGGCGGGGCACAGGAAGAC[A/G]AAAGGATGCCCCAGGCCCCAGATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604178 MIM: 601843 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPL18A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RPL18A - ribosomal protein L18a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000980.3 | Intron | NP_000971.1 |
SLC5A5 - solute carrier family 5 member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORA68 - small nucleolar RNA, H/ACA box 68 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |