Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTATTAGATTTCACAAAACAGAAA[A/G]AATTCTGAGCCAGGCACAATGGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUGGC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NUGGC - nuclear GTPase, germinal center associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010906.1 | 2044 | UTR 3 | NP_001010906.1 | |||
XM_011544523.2 | 2044 | Intron | XP_011542825.1 | |||
XM_011544524.2 | 2044 | Intron | XP_011542826.1 | |||
XM_011544525.1 | 2044 | UTR 3 | XP_011542827.1 | |||
XM_011544526.2 | 2044 | Intron | XP_011542828.1 | |||
XM_017013403.1 | 2044 | Intron | XP_016868892.1 |