Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300485 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCOR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BCOR - BCL6 corepressor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001123383.1 | Intron | NP_001116855.1 | ||||
NM_001123384.1 | Intron | NP_001116856.1 | ||||
NM_001123385.1 | Intron | NP_001116857.1 | ||||
NM_017745.5 | Intron | NP_060215.4 | ||||
XM_005272616.1 | Intron | XP_005272673.1 | ||||
XM_005272618.3 | Intron | XP_005272675.1 | ||||
XM_005272619.3 | Intron | XP_005272676.1 | ||||
XM_005272620.3 | Intron | XP_005272677.1 | ||||
XM_006724536.3 | Intron | XP_006724599.1 | ||||
XM_011543929.2 | Intron | XP_011542231.1 | ||||
XM_011543930.1 | Intron | XP_011542232.1 | ||||
XM_011543931.2 | Intron | XP_011542233.1 | ||||
XM_017029615.1 | Intron | XP_016885104.1 | ||||
XM_017029616.1 | Intron | XP_016885105.1 |