Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
61 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120280 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL11A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COL11A1 - collagen type XI alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190709.1 | 6922 | UTR 3 | NP_001177638.1 | |||
NM_001854.3 | 6922 | UTR 3 | NP_001845.3 | |||
NM_080629.2 | 6922 | UTR 3 | NP_542196.2 | |||
NM_080630.3 | 6922 | UTR 3 | NP_542197.3 | |||
XM_017000334.1 | 6922 | UTR 3 | XP_016855823.1 | |||
XM_017000335.1 | 6922 | Intron | XP_016855824.1 | |||
XM_017000336.1 | 6922 | Intron | XP_016855825.1 | |||
XM_017000337.1 | 6922 | Intron | XP_016855826.1 |
Set Membership: |
HapMap |