Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATTTTGAGCAGGACTATGGTTAA[A/G]CCCTTCCAGAAACCTGATAATTGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611145 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC30A8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SLC30A8 - solute carrier family 30 member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172811.1 | Intron | NP_001166282.1 | ||||
NM_001172813.1 | Intron | NP_001166284.1 | ||||
NM_001172814.1 | Intron | NP_001166285.1 | ||||
NM_001172815.2 | Intron | NP_001166286.1 | ||||
NM_173851.2 | Intron | NP_776250.2 |
Set Membership: |
HapMap |