Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607048 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STARD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
STARD3 - StAR related lipid transfer domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165937.1 | Intron | NP_001159409.1 | ||||
NM_001165938.1 | Intron | NP_001159410.1 | ||||
NM_006804.3 | Intron | NP_006795.3 | ||||
XM_006721646.2 | Intron | XP_006721709.1 | ||||
XM_011524210.1 | Intron | XP_011522512.1 | ||||
XM_011524211.2 | Intron | XP_011522513.1 | ||||
XM_011524213.1 | Intron | XP_011522515.1 | ||||
XM_017024041.1 | Intron | XP_016879530.1 |
Set Membership: |
HapMap |