Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAATTGCAGTGTGAGCTTATACAA[A/T]TGTACTACATATTTTTATACATCTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602941 MIM: 118890 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCAR1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BCAR1 - BCAR1, Cas family scaffolding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CTRB1 - chymotrypsinogen B1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100506281 - uncharacterized LOC100506281 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |