Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTACTATTTGCATTTTATTGTCTG[A/G]ATACCAAACTTATGCCTGGCTTTAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 185605 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYT1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SYT1 - synaptotagmin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135805.1 | Intron | NP_001129277.1 | ||||
NM_001135806.1 | Intron | NP_001129278.1 | ||||
NM_001291901.1 | Intron | NP_001278830.1 | ||||
NM_005639.2 | Intron | NP_005630.1 | ||||
XM_005269113.2 | Intron | XP_005269170.1 | ||||
XM_006719576.1 | Intron | XP_006719639.1 | ||||
XM_011538710.1 | Intron | XP_011537012.1 | ||||
XM_017019909.1 | Intron | XP_016875398.1 |
Set Membership: |
HapMap |