Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAACAGCAGCTTCCATATCCTGAA[C/T]GCCTCTAATGTGCCAGGCATGTGTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600378 MIM: 614918 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IMMT PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IMMT - inner membrane mitochondrial protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100169.1 | Intron | NP_001093639.1 | ||||
NM_001100170.1 | Intron | NP_001093640.1 | ||||
NM_006839.2 | Intron | NP_006830.2 | ||||
XM_005264110.3 | Intron | XP_005264167.1 | ||||
XM_005264113.1 | Intron | XP_005264170.1 | ||||
XM_005264114.1 | Intron | XP_005264171.1 | ||||
XM_011532497.2 | Intron | XP_011530799.1 | ||||
XM_017003194.1 | Intron | XP_016858683.1 | ||||
XM_017003195.1 | Intron | XP_016858684.1 |
PTCD3 - pentatricopeptide repeat domain 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |