Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTGGGAAATAATGTAGAATTAAG[A/C]TCTCAAATATACTATGCAATTGTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601677 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFA5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NDUFA5 - NADH:ubiquinone oxidoreductase subunit A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282419.2 | Intron | NP_001269348.1 | ||||
NM_001282420.2 | Intron | NP_001269349.1 | ||||
NM_001282421.2 | Intron | NP_001269350.1 | ||||
NM_001282422.2 | Intron | NP_001269351.1 | ||||
NM_001291304.1 | Intron | NP_001278233.1 | ||||
NM_005000.4 | Intron | NP_004991.1 |
Set Membership: |
HapMap |