Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCTGAACCCACAGGCCTCCCAT[A/G]TATGCCCTCCCCCCAGGGTGCCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138032 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLP1R PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLP1R - glucagon like peptide 1 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002062.4 | 660 | Intron | NP_002053.3 | |||
XM_017010750.1 | 660 | Missense Mutation | TAT,TGT | Y,C 26 | XP_016866239.1 | |
XM_017010751.1 | 660 | Missense Mutation | TAT,TGT | Y,C 26 | XP_016866240.1 |