Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608335 MIM: 610035 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHO1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PLEKHO1 - pleckstrin homology domain containing O1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304722.1 | 115 | Intron | NP_001291651.1 | |||
NM_001304723.1 | 115 | UTR 5 | NP_001291652.1 | |||
NM_001304724.1 | 115 | Intron | NP_001291653.1 | |||
NM_016274.5 | 115 | Intron | NP_057358.2 | |||
XM_017001420.1 | 115 | Intron | XP_016856909.1 |
VPS45 - vacuolar protein sorting 45 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |