Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTTAGGTAAGCATACATTAAAG[G/T]TTTCGAGATATTATCTCCTCATTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603008 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDH8 - cadherin 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001796.4 | 5362 | Intron | NP_001787.2 | |||
XM_005255760.3 | 5362 | UTR 3 | XP_005255817.1 | |||
XM_017022829.1 | 5362 | Intron | XP_016878318.1 |
Set Membership: |
HapMap |