Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138440 MIM: 182465 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GART PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GART - phosphoribosylglycinamide formyltransferase, phosphoribosylglycinamide synthetase, phosphoribosylaminoimidazole synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000819.4 | Intron | NP_000810.1 | ||||
NM_001136005.1 | Intron | NP_001129477.1 | ||||
NM_001136006.1 | Intron | NP_001129478.1 | ||||
NM_175085.2 | Intron | NP_780294.1 | ||||
XM_005260941.1 | Intron | XP_005260998.1 | ||||
XM_006723989.1 | Intron | XP_006724052.1 | ||||
XM_006723990.1 | Intron | XP_006724053.1 | ||||
XM_011529526.1 | Intron | XP_011527828.1 |
MIR6501 - microRNA 6501 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SON - SON DNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291411.1 | Intron | NP_001278340.1 | ||||
NM_001291412.1 | Intron | NP_001278341.1 | ||||
NM_032195.2 | Intron | NP_115571.2 | ||||
NM_138927.2 | Intron | NP_620305.2 |
Set Membership: |
HapMap |