Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300311 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TEX11 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TEX11 - testis expressed 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003811.1 | Intron | NP_001003811.1 | ||||
NM_031276.2 | Intron | NP_112566.2 | ||||
XM_011530994.1 | Intron | XP_011529296.1 | ||||
XM_011530996.1 | Intron | XP_011529298.1 | ||||
XM_017029649.1 | Intron | XP_016885138.1 | ||||
XM_017029650.1 | Intron | XP_016885139.1 | ||||
XM_017029651.1 | Intron | XP_016885140.1 | ||||
XM_017029652.1 | Intron | XP_016885141.1 |