Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTAAATGGGCAGTGTGTGGTCAGT[A/G]GAATGGCAGTTTGAGAGCTATGTAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608155 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYNPO PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SYNPO - synaptopodin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001109974.2 | Intron | NP_001103444.1 | ||||
NM_001166208.1 | Intron | NP_001159680.1 | ||||
NM_001166209.1 | Intron | NP_001159681.1 | ||||
NM_007286.5 | Intron | NP_009217.3 | ||||
XM_005268370.1 | Intron | XP_005268427.1 | ||||
XM_005268371.1 | Intron | XP_005268428.1 | ||||
XM_006714755.3 | Intron | XP_006714818.1 | ||||
XM_011537552.2 | Intron | XP_011535854.1 | ||||
XM_017009007.1 | Intron | XP_016864496.1 | ||||
XM_017009008.1 | Intron | XP_016864497.1 |
Set Membership: |
HapMap |