Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611393 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM110A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM110A - family with sequence similarity 110 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042353.2 | 563 | Intron | NP_001035812.1 | |||
NM_001289145.1 | 563 | Intron | NP_001276074.1 | |||
NM_001289146.1 | 563 | Intron | NP_001276075.1 | |||
NM_001289147.1 | 563 | Intron | NP_001276076.1 | |||
NM_031424.5 | 563 | Intron | NP_113612.1 | |||
NM_207121.4 | 563 | Intron | NP_997004.1 | |||
XM_011529381.2 | 563 | UTR 5 | XP_011527683.1 |